Order Entry
Italy
ContactUsLinkComponent
Anti-Staphylococcus sp. Mouse Monoclonal Antibody [clone: 17-5]
Anti-Staphylococcus sp. Mouse Monoclonal Antibody [clone: 17-5]
Catalog # USBIS7965-80
Supplier:  US Biological
Anti-Staphylococcus sp. Mouse Monoclonal Antibody [clone: 17-5]
Catalog # USBIS7965-80
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Staphylococcus sp.
  • Clonality:
    Monoclonal
  • Clone:
    17-5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG3
  • Format:
    Supernatant
  • Form:
    liquid
  • Storage buffer:
    0,02% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~200 ug/ml
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: Staphylococcus sp.
Clonality: Monoclonal
Clone: 17-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: