Anti-Specific Intracellular Adhesion Molecule Grabbing Nonintegrin R1 Rat Monoclonal Antibody [clone: ER-TR9]
Catalog # USBIS5370-90
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Specific Intracellular Adhesion Molecule Grabbing Nonintegrin R1
- Clonality:Monoclonal
- Clone:ER-TR9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgM
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:50; Immunochemistry: 1:50
Type: Primary
Antigen: Specific Intracellular Adhesion Molecule Grabbing Nonintegrin R1
Clonality: Monoclonal
Clone: ER-TR9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: