Anti-SIRP alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9A204]
Catalog # USBIS1013-86ZB3
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:SIRP alpha
- Clonality:Monoclonal
- Clone:9A204
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: SIRP alpha
Clonality: Monoclonal
Clone: 9A204
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human