Anti-PTPN11 Mouse Monoclonal Antibody [clone: SIT-01]
USBIS1013-30D
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase, non-receptor type 11
- Antigen symbol:PTPN11
- Clonality:Monoclonal
- Clone:SIT-01
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 15mM sodium azide, pH 7,4.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: Monoclonal
Clone: SIT-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse