Anti-SETD8 Rabbit Polyclonal Antibody
200 µl
Catalog # USBIS1012-28C
Supplier: US Biological
Jump to:
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: SETD8 (SET domain containing (lysine methyltransferase) 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Documents
Specifications
- Size:200 µl
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by Protein G chromatography
- Antibody type:Primary
- Epitope:N-terminal
- Form:liquid
- Isotype:IgG
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:SET domain containing (lysine methyltransferase) 8
- Cat. No.:USBIS1012-28C
- Reactivity:Human|Mouse
- Conjugation:Unconjugated
- Antigen symbol:SETD8
- Storage buffer:PBS, 0,09% sodium azide.
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



