Order Entry
Italy
ContactUsLinkComponent
Anti-RSK1, RSK2 Rabbit Polyclonal Antibody
Anti-RSK1, RSK2 Rabbit Polyclonal Antibody
Catalog # USBIR9399-15C
Supplier:  US Biological
Anti-RSK1, RSK2 Rabbit Polyclonal Antibody
Catalog # USBIR9399-15C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    RSK1, RSK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Chicken,
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Ser380, phospho-Ser386
  • Storage buffer:
    PBS(without Mg2+ and Ca2+), 1mg/ml BSA (IgG, protease free), pH 7,2, 0,1% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    10 Pieces
  • Pk:
    10

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: RSK1, RSK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse