Anti-RSK1, RSK2 Rabbit Polyclonal Antibody
Codice catalogo USBIR9399-15B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:RSK1, RSK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Chicken,Human
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser363, phospho-Ser369
- Storage buffer:PBS(without Mg2+ and Ca2+), pH 7,3, 1mg/mlBSA (IgG, protease free), 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:10 Pieces
- Pk:10
Specifications
About this item
Recommended dilution: Western Blot: 1:1000
Type: Primary
Antigen: RSK1, RSK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human