Order Entry
Italy
ContactUsLinkComponent
Anti-RNA 5.8s, Ribosomal Mouse Monoclonal Antibody [clone: 9C93]
Anti-RNA 5.8s, Ribosomal Mouse Monoclonal Antibody [clone: 9C93]
Catalog # USBIR2079
Supplier:  US Biological
Anti-RNA 5.8s, Ribosomal Mouse Monoclonal Antibody [clone: 9C93]
Catalog # USBIR2079
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    RNA 5.8s, Ribosomal
  • Clonality:
    Monoclonal
  • Clone:
    9C93
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG3 kappa
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen: RNA 5.8s, Ribosomal
Clonality: monoclonal
Clone: 9C93
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: