Anti-RIPK3 Rabbit Polyclonal Antibody
Catalog # USBIR2031-74J
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 3
- Antigen symbol:RIPK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:480 to 518
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifiche
Informazioni su questo articolo
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: RIPK3 (receptor-interacting serine-threonine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human