Order Entry
Italy
ContactUsLinkComponent
 
Anti-QRFP Pyroglutamylated RFamide Peptide 43 Rabbit Polyclonal Antibody (Carboxyfluorescein)
  USBIQ0575-50C
Anti-QRFP Pyroglutamylated RFamide Peptide 43 Rabbit Polyclonal Antibody (Carboxyfluorescein)
  USBIQ0575-50C

 

  • Antibody type:
    Primary
  • Antigen symbol:
    QRFP Pyroglutamylated RFamide Peptide 43
  • Clonality:
    Polyclonal
  • Conjugation:
    FAM (Carboxyfluorescein)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    liquid
  • Amino acid number:
    154 to 196
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: QRFP Pyroglutamylated RFamide Peptide 43
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human