Anti-PP6C Rabbit Polyclonal Antibody
USBIP9107-56
- Antibody type:Primary
- Antigen symbol:PP6C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,08% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:100; Immunoprecipitation: 10-20 ?g/ml
Type: Primary
Antigen: PP6C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat