Anti-Prokineticin 2-long Rabbit polyclonal antibody Carboxyfluorescein
Catalog # USBIP9008-84C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Prokineticin 2-long
- Clonality:Polyclonal
- Conjugation:FAM (Carboxyfluorescein)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Amino acid number:48 to 68
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Prokineticin 2-long
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human