Order Entry
Italy
ContactUsLinkComponent
Anti-Polyubiquitin, Lysine 48-Linkage Specific Rabbit Polyclonal Antibody
Anti-Polyubiquitin, Lysine 48-Linkage Specific Rabbit Polyclonal Antibody
Catalog # USBIP4480-48
Supplier:  US Biological
Anti-Polyubiquitin, Lysine 48-Linkage Specific Rabbit Polyclonal Antibody
Catalog # USBIP4480-48
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Polyubiquitin, Lysine 48-Linkage Specific
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish,
    Mouse,
    Hamster,
    Pig,
    Monkey,
    Chicken,
    C. elegans,
    Human,
    Bovine,
    Xenopus laevis,
    Dog,
    Rat
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    10mM HEPES, pH 7,5, 150mM sodium chloride,0,1mg/ml BSA, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Polyubiquitin, Lysine 48-Linkage Specific
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, C. elegans, Chicken, Dog, Hamster, Human, Monkey, Mouse, Pig, Rat, Xenopus laevis, Zebrafish