Anti-Pix, beta Rabbit Polyclonal Antibody (Biotin)
Catalog # USBIP4210-59B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:PIX beta
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: ELISA: 1:5000-1:10000); Western Blot: 1:5000-1:10000
Type: Primary
Antigen: Pix, beta
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat