Anti-Peptide Histidine Methioninamide 27 Rabbit Polyclonal Antibody
Catalog # USBIP3282-04V
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Peptide Histidine Methioninamide 27
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Format:Serum
- Form:lyophilized powder
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500
Type: Primary
Antigen: Peptide Histidine Methioninamide 27
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig