Order Entry
Italy
ContactUsLinkComponent
Anti-PAEL Receptor 37 Like 1 Mouse Monoclonal Antibody [clone: 8B39]
Anti-PAEL Receptor 37 Like 1 Mouse Monoclonal Antibody [clone: 8B39]
  USBIP1799-03
Anti-PAEL Receptor 37 Like 1 Mouse Monoclonal Antibody [clone: 8B39]
  USBIP1799-03

 

  • Antibody type:
    Primary
  • Antigen name:
    PAEL Receptor 37 Like 1
  • Clonality:
    Monoclonal
  • Clone:
    8B39
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, 5% trehalose.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml (after reconstitution)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: PAEL Receptor 37 Like 1
Clonality: monoclonal
Clone: 8B39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human