Order Entry
Italy
ContactUsLinkComponent
Anti-PACAP-27 Rabbit Polyclonal Antibody
Anti-PACAP-27 Rabbit Polyclonal Antibody
Catalog # USBIP1775-02A
Supplier:  US Biological
Anti-PACAP-27 Rabbit Polyclonal Antibody
Catalog # USBIP1775-02A
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PACAP27
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Sheep,
    Human,
    Rat
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Antigen synonyms:
    PACAP-27
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500

Type: Primary
Antigen: PACAP-27
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Sheep