Anti-OAS2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBIO2970-05-PE
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:2'-5'-oligoadenylate synthetase 2, 69/71kDa
- Antigen symbol:OAS2
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Epitope:C-terminal
- Form:liquid
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein G chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: OAS2 (2'-5'-oligoadenylate synthetase 2, 69/71kDa)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human