Anti-Nudix-type Motif 21 Mouse Monoclonal Antibody [clone: 8C255]
USBIN7400-25
- Antibody type:Primary
- Antigen name:Nudix-type Motif 21
- Clonality:Monoclonal
- Clone:8C255
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Flow Cytometry: 0.5-2?g/samplr; Western Blot: 0.2-2?g/ml; Immunoprecipitation: 100-500 ?g/sample; Immunochemistry: 2-100 ?g/ml
Type: Primary
Antigen: Nudix-type Motif 21
Clonality: monoclonal
Clone: 8C255
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat