Anti-Nuclear Receptor Rev-ErbA alpha Rabbit Polyclonal Antibody
Catalog # USBIN6889-85G
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Rev-ErbA alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:Tris-glycine, pH 7,4, 0,15mM sodium chloride, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: Nuclear Receptor Rev-ErbA alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human