Anti-NOVA 2 RNA Binding Protein Rabbit Polyclonal Antibody
Catalog # USBIN5376-05-50
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:NOVA 2 RNA Binding Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rodent,Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NOVA 2 RNA Binding Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rodent