Order Entry
Italy
ContactUsLinkComponent
 
Anti-Nicotinic Receptor, beta 4 Rabbit Polyclonal Antibody
  USBIN2565-33E
Anti-Nicotinic Receptor, beta 4 Rabbit Polyclonal Antibody
  USBIN2565-33E

 

  • Antibody type:
    Primary
  • Antigen name:
    Nicotinic acetylcholine receptor beta 4
  • Antigen symbol:
    CHRNB4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Amino acid number:
    490 to 498
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by precipitation, dialysis and chromatography
  • Size:
    5 ml
  • Pk:
    5 mL

 

 

Type: Primary
Antigen: Nicotinic Receptor, beta 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human