Order Entry
Italy
ContactUsLinkComponent
Anti-Nerve Growth Factor Receptor, Human Mouse Monoclonal Antibody [clone: 1.A.31]
Anti-Nerve Growth Factor Receptor, Human Mouse Monoclonal Antibody [clone: 1.A.31]
Catalog # USBIN2050-15-250
Supplier:  US Biological
Anti-Nerve Growth Factor Receptor, Human Mouse Monoclonal Antibody [clone: 1.A.31]
Catalog # USBIN2050-15-250
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nerve Growth Factor Receptor, Human
  • Clonality:
    Monoclonal
  • Clone:
    1.A.31
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Type: Primary
Antigen: Nerve Growth Factor Receptor, Human
Clonality: Monoclonal
Clone: 1.A.31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human