Order Entry
Italy
ContactUsLinkComponent
Anti-Mucin 2 Mouse Monoclonal Antibody [clone: 5F270]
Anti-Mucin 2 Mouse Monoclonal Antibody [clone: 5F270]
  USBIM4700-24
Anti-Mucin 2 Mouse Monoclonal Antibody [clone: 5F270]
  USBIM4700-24

 

  • Antibody type:
    Primary
  • Antigen name:
    Mucin 2
  • Antigen symbol:
    MUC2
  • Clonality:
    Monoclonal
  • Clone:
    5F270
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Format:
    Cell Culture supernatant
  • Form:
    liquid
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Recommended dilution: Immunochemistry: 1:10-1:40

Type: Primary
Antigen: Mucin 2
Clonality: monoclonal
Clone: 5F270
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human