Anti-MHC Class 2 Mouse Monoclonal Antibody [clone: 53-5D2]
Catalog # USBIM3887-09C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 2
- Clonality:Monoclonal
- Clone:53-5D2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Form:liquid
- Storage buffer:PBS, 0,01% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Recommended dilution: Flow Cytometry: 1:10; Immunochemistry: 1:100
Type: Primary
Antigen: MHC Class 2
Clonality: monoclonal
Clone: 53-5D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat