Anti-MHC class I, H-2Kb, Db Mouse Monoclonal Antibody
Catalog # USBIM3885-80G-500
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H-2Kb, Db
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: MHC class I, H-2Kb, Db
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse