Anti-MHC Class 1, H2, Dk Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15-5-5.3]
Catalog # USBIM3885-60H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H2, Dk
- Clonality:Monoclonal
- Clone:15-5-5.3
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1, H2, Dk
Clonality: monoclonal
Clone: 15-5-5.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse