Anti-MHC Class 1, H2, Db, alpha3 Mouse Monoclonal Antibody [clone: 28-14-8]
Catalog # USBIM3885-59A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1, H2, Db, alpha3
- Clonality:Monoclonal
- Clone:28-14-8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:liquid
- Storage buffer:100mM borate buffered saline, pH 8.0. No oramine- containing buffer salts .
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1, H2, Db, alpha3
Clonality: monoclonal
Clone: 28-14-8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse