Order Entry
Italy
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Catalog # USBIM2865-11M
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Polyclonal Antibody
Catalog # USBIM2865-11M
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Monkey,
    Chicken,
    Human,
    Xenopus laevis,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Thr202, phospho-Tyr204
  • Storage buffer:
    10mM HEPES, pH 7,5, 0,01% BSA, 0,09% BSA and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: Western Blot:1:1000

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Dog, Human, Monkey, Mouse, Rat, Xenopus laevis