Order Entry
Italy
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 8A17]
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 8A17]
Catalog # USBIM2352-20C
Supplier:  US Biological
Anti-MAP Kinase p44/42 Rabbit Monoclonal Antibody [clone: 8A17]
Catalog # USBIM2352-20C
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Monoclonal
  • Clone:
    8A17
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish,
    Hamster,
    Drosophila,
    Monkey,
    C. elegans,
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Modification:
    phospho-Thr202, phospho-Tyr204
  • Storage buffer:
    10mMsodium HEPES (pH 7,5), 150mM NaCl, 100ug/ml BSA and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:200; Flow Cytometry: 1:100; Western Blot: 1:2000; Immunochemistry: 1:200; Immunoprecipitation: 1:50

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: monoclonal
Clone: 8A17
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, C. elegans, Drosophila, Hamster, Human, Monkey, Mouse, Rat, Zebrafish