Order Entry
Italy
ContactUsLinkComponent
Anti-MAP Kinase p44/42 Mouse Monoclonal Antibody [clone: 12E12]
Anti-MAP Kinase p44/42 Mouse Monoclonal Antibody [clone: 12E12]
Catalog # USBIM2352-10E
Supplier:  US Biological
Anti-MAP Kinase p44/42 Mouse Monoclonal Antibody [clone: 12E12]
Catalog # USBIM2352-10E
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MAP Kinase p44/42
  • Clonality:
    Monoclonal
  • Clone:
    12E12
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1:2000-1:5000

Type: Primary
Antigen: MAP Kinase p44/42
Clonality: monoclonal
Clone: 12000000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat