Order Entry
Italy
ContactUsLinkComponent
Anti-MAP Kinase 14 Rabbit Polyclonal Antibody
Anti-MAP Kinase 14 Rabbit Polyclonal Antibody
Catalog # USBIM2352-03B
Supplier:  US Biological
Anti-MAP Kinase 14 Rabbit Polyclonal Antibody
Catalog # USBIM2352-03B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 14
  • Antigen symbol:
    MAPK14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Modification:
    phospho-Thr180
  • Storage buffer:
    PBS(without Mg2+ and Ca2+), pH 7,4, 150mMsodium chloride, 0,02% sodium azide, 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry:1:50-1:100

Type: Primary
Antigen: MAP Kinase 14
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat