Anti-Ly49C,F,H,I Hamster Monoclonal Antibody (FITC (Fluorescein)) [clone: 14B11]
USBIL7732-01A
- Antibody type:Primary
- Antigen symbol:LY49C,F,H,I
- Clonality:Monoclonal
- Clone:14B11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: Ly49C,F,H,I
Clonality: monoclonal
Clone: 14B11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse