Anti-LCN2 Mouse Monoclonal Antibody [clone: 048-17]
USBIL2499-17G
- Antibody type:Primary
- Antigen name:lipocalin 2
- Antigen symbol:LCN2
- Clonality:Monoclonal
- Clone:048-17
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Pig
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 048-17
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Pig