Anti-LAT Mouse Monoclonal Antibody [clone: LAT.10-17 (10-17)]
USBIL1390-01K
- Antibody type:Primary
- Antigen name:linker for activation of T cells
- Antigen symbol:LAT
- Clonality:Monoclonal
- Clone:LAT.10-17 (10-17)
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: monoclonal
Clone: LAT.10-17 (10-17)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human