Order Entry
Italy
ContactUsLinkComponent
Anti-KRT17 Mouse Monoclonal Antibody [clone: Ks 17.E3]
Anti-KRT17 Mouse Monoclonal Antibody [clone: Ks 17.E3]
Catalog # USBIK0199-20D
Supplier:  US Biological
Anti-KRT17 Mouse Monoclonal Antibody [clone: Ks 17.E3]
Catalog # USBIK0199-20D
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 17
  • Antigen symbol:
    KRT17
  • Clonality:
    Monoclonal
  • Clone:
    Ks 17.E3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    lyophilized powder
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    1 ml
  • Pk:
    1 mL

Specifications

About this item

Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: KRT17 (keratin 17)
Clonality: monoclonal
Clone: Ks 17.E3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat