Order Entry
Italy
ContactUsLinkComponent
Anti-Keratin 8, 18, 19 Mouse Monoclonal Antibody [clone: 2A4]
Anti-Keratin 8, 18, 19 Mouse Monoclonal Antibody [clone: 2A4]
  USBIK0199-08X
undefined
Anti-Keratin 8, 18, 19 Mouse Monoclonal Antibody [clone: 2A4]
  USBIK0199-08X

 

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 8, 18,19
  • Antigen symbol:
    KRT8, 18,19
  • Clonality:
    Monoclonal
  • Clone:
    2A4
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Pig
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 1% BSA, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Fast protein liquid chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Recommended dilution: Western Blot: 1:1000-1:4000; Immunochemistry: 1:100-200

Type: Primary
Antigen: Keratin 8, 18,19
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig