Order Entry
Italy
ContactUsLinkComponent
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Catalog # USBIK0199-08W
Supplier:  US Biological
undefined
Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
Catalog # USBIK0199-08W
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    keratin 8
  • Antigen symbol:
    KRT8
  • Clonality:
    Monoclonal
  • Clone:
    C-51
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 15mM sodium azide, pH 7,4.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: C-51
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human