Anti-KRT8 Mouse Monoclonal Antibody [clone: C-43]
Catalog # USBIK0199-08H
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:keratin 8
- Antigen symbol:KRT8
- Clonality:Monoclonal
- Clone:C-43
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Rabbit,Sheep,Human,Bovine
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 15mM sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: C-43
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig, Rabbit, Sheep