Anti-Jumonji/ARID Domain Containing Protein 2 Goat Polyclonal Antibody
Catalog # USBIJ7876-21
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Jumonji/ARID Domain Containing Protein 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-HRSAQDLRKQVSKVN
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by peptide affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1: 32000
Type: Primary
Antigen: Jumonji/ARID Domain Containing Protein 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human