Anti-IDH1, IDH2 Rabbit Polyclonal Antibody
Catalog # USBII8850-06
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:IDH1, IDH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Recommended dilution: ELISA: 1:400-1:1600; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: IDH1, IDH2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig