Order Entry
Italy
ContactUsLinkComponent
Anti-Interleukin 34, IN Rabbit Polyclonal Antibody
Anti-Interleukin 34, IN Rabbit Polyclonal Antibody
Catalog # USBII8443-93B
Supplier:  US Biological
Anti-Interleukin 34, IN Rabbit Polyclonal Antibody
Catalog # USBII8443-93B
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 34, IN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: Interleukin 34, IN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat