Anti-Interleukin 18 Mouse Monoclonal Antibody (Biotin) [clone: 3H2456]
Catalog # USBII8439-95C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 18
- Antigen symbol:IL18
- Clonality:Monoclonal
- Clone:3H2456
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,09% sodium azide
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:250 µg
- Pk:250 µG
Specifications
About this item
Type: Primary
Antigen: Interleukin 18
Clonality: monoclonal
Clone: 3H2456
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig