Anti-Interleukin-17 Receptor A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K116]
USBII8439-35H
- Antibody type:Primary
- Antigen name:Interleukin-17 Receptor A
- Clonality:Monoclonal
- Clone:12K116
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:liquid
- Amino acid number:32 to 322
- Storage buffer:saline, 0,5% BSA, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,025 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Do not freeze!
Type: Primary
Antigen: Interleukin-17 Receptor A
Clonality: monoclonal
Clone: 12K116
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse