Order Entry
Italy
ContactUsLinkComponent
Anti-Interleukin 17 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H34]
Anti-Interleukin 17 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H34]
Catalog # USBII8439-08H
Supplier:  US Biological
Anti-Interleukin 17 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H34]
Catalog # USBII8439-08H
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 17
  • Antigen symbol:
    IL17
  • Clonality:
    Monoclonal
  • Clone:
    7H34
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    liquid
  • Amino acid number:
    20 to 155
  • Storage buffer:
    PBS, 0,1% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,025 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Do not freeze!

Type: Primary
Antigen: Interleukin 17
Clonality: monoclonal
Clone: 7H34
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human