Anti-Interleukin 17 Rat Monoclonal Antibody [clone: 10B2376]
Catalog # USBII8439-04L
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17
- Antigen symbol:IL17
- Clonality:Monoclonal
- Clone:10B2376
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Form:liquid
- Storage buffer:borate buffered saline, pH 8,2
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
Type: Primary
Antigen: Interleukin 17
Clonality: monoclonal
Clone: 10B2376
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse