Anti-IL13 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B2375]
Catalog # USBII8435-02P
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 13
- Antigen symbol:IL13
- Clonality:Monoclonal
- Clone:10B2375
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:liquid
- Storage buffer:Tris-sodium chloride, pH 7,2, 1% BSA, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ion exchange chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Recommended dilution: Flow Cytometry:
Type: Primary
Antigen: IL13 (interleukin 13)
Clonality: monoclonal
Clone: 10B2375
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human