Anti-Interleukin 12 beta Mouse Monoclonal Antibody [clone: 9G467]
Catalog # USBII8434-27C
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 12 Beta
- Antigen symbol:IL12B
- Clonality:Monoclonal
- Clone:9G467
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-2000
Type: Primary
Antigen: Interleukin 12 beta
Clonality: monoclonal
Clone: 9G467
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human