Order Entry
Italy
ContactUsLinkComponent
 
Anti-IL8 Mouse Monoclonal Antibody [clone: 5D21]
  USBII8430-06A
Anti-IL8 Mouse Monoclonal Antibody [clone: 5D21]
  USBII8430-06A

 

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 8
  • Antigen symbol:
    IL8
  • Clonality:
    Monoclonal
  • Clone:
    5D21
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

Recommended dilution: ELISA: 1:10000

Type: Primary
Antigen: IL8 (interleukin 8)
Clonality: monoclonal
Clone: 5D21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human