Anti-Interferon alpha 1 Mouse Monoclonal Antibody [clone: 2-52]
Catalog # USBII7661-60B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Interferon alpha 1
- Antigen symbol:IFNA1
- Clonality:Monoclonal
- Clone:2-52
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, 0,05% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Interferon alpha 1
Clonality: monoclonal
Clone: 19025
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human